Learn More
Kern Sohn Compact Laboratory Balance PFB
Click to view available options
Capacity:
120 g
200 g
300 g
600 g
1200 g
2000 g
3000 g
6000 g
Readability:
0.001 g
0.01 g
0.05 g
0.1 g
Unit Size:
Each
Description
- Easy to use: All primary functions have their own key on the keypad
- Compact size, practical for small spaces
- Capacity display: A bar graph display lights up to show how much of the weighing range is still available
- Level indicator and levelling feet for precise levelling of the scale, fitted as standard, to give the most accurate weighing result
Specifications
Specifications
| Capacity | 6000 g |
| Readability | 0.05 g |
| Linearity | 0.15 g |
| Type | Compact Precision Balance |
| Length (Metric) Pan | 190 mm |
| Width (Metric) Pan | 180 mm |
| Electrical Requirements | Universal Mains Adapter |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.