Learn More
BRAND™ Compact Burette, BLAUBRAND™, Class AS, DE-M, with Stopcock
Shop All Brand Products
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Unit Size:
Each
Descrizione
- With removable PTFE stopcock, fast to dismantle and easy to clean
- Simple repair - all individual components replaceable
- To DIN EN ISO 385
- Calibrated to deliver (TD, Ex)
All BLAUBRAND™ burettes are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate.
Specifica
Specifica
| Color | Amber |
| Capacity (Metric) | 25 mL |
| Material | Borosilicate Glass (3.3) |
| Graduations | 0.05 mL |
| Includes | Burette tube, stopcock with precision tip, one batch certificate |
| Stopcock Material | PTFE |
| Type | Compact Buret |
| Class | Class AS |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto