Learn More
Emma Safety Footwear Comodius Safety Shoes
Click to view available options
Size:
39
40
41
42
43
44
46
47
Unit Size:
1 pair
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- Vibram™ Nitrile Rubber Heat Resistant Outsole
Specifications
Specifications
| Certifications/Compliance | EN ISO 20345:2011 |
| Closure Type | Laces |
| Color | Black |
| Design | Comodius |
| Liner Material | Full grain Nappa leather |
| Material | Leather |
| Size | 44 |
| Toe Material | Aluminum |