Learn More
Abeba™ Comfort 6874 Shoes
Stylish hand-sewn seams and PU sole with high slip resistance.
Brand: Abeba™ 6874/42
Additional Details : Weight : 0.48000kg
Description
- CE EN ISO 20347:2012 OB A, E SRB
- Smooth leather white
- Slip resistance PU outsole sole
- Replaceable Reflexor insole with nubs (article no. 3572)
- Sanitized treated
- Antistatic
- Heel strap non-foldable, adjustable
- Lining microfiber, respiration absorbing
- Instep area with elastic band
- Hand-sewn seam
Specifications
CE EN ISO 20347:2012 OB A, E SRB | |
Microfiber | |
42 | |
Polyurethane |
White | |
Leather | |
Slip-resistant | |
6874 |