Learn More
Abeba™ Comfort 6874 Shoes
Stylish hand-sewn seams and PU sole with high slip resistance.
94.98€
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E SRB |
---|---|
Color | White |
Liner Material | Microfiber |
Material | Leather |
Sole Features | Slip-resistant |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15692348
|
Abeba™
6874/36 |
36 |
94.98€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15602358
|
Abeba™
6874/37 |
37 |
94.98€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15612358
|
Abeba™
6874/38 |
38 |
94.98€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15622358
|
Abeba™
6874/39 |
39 |
94.98€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15632358
|
Abeba™
6874/40 |
40 |
94.98€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15642358
|
Abeba™
6874/41 |
41 |
94.98€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15652358
|
Abeba™
6874/42 |
42 |
94.98€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15662358
|
Abeba™
6874/43 |
43 |
94.98€
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, E SRB
- Smooth leather white
- Slip resistance PU outsole sole
- Replaceable Reflexor insole with nubs (article no. 3572)
- Sanitized treated
- Antistatic
- Heel strap non-foldable, adjustable
- Lining microfiber, respiration absorbing
- Instep area with elastic band
- Hand-sewn seam
Specifications
CE EN ISO 20347:2012 OB A, E SRB | |
Microfiber | |
Slip-resistant | |
6874 |
White | |
Leather | |
Polyurethane |