Learn More
Abeba™ Comfort 6874 Shoes
Stylish hand-sewn seams and PU sole with high slip resistance.
Brand:  Abeba™ 6874/40
Additional Details : Weight : 0.48000kg
 View more versions of this product
Product Code. 15632358
  • 94.98€ / 1 pair
Estimated Shipment: 04-06-2024
Description
  • CE EN ISO 20347:2012 OB A, E SRB
  • Smooth leather white
  • Slip resistance PU outsole sole
  • Replaceable Reflexor insole with nubs (article no. 3572)
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Lining microfiber, respiration absorbing
  • Instep area with elastic band
  • Hand-sewn seam
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRB
Microfiber
40
Polyurethane
White
Leather
Slip-resistant
6874