Learn More
Fisherbrand™ Combs for TVG-SYS Vari-gel MIDI Horizontal Gel Electrophoresis Unit
Product Code. 11796185
Click to view available options
No. of Wells:
8
12
15
20
Volume (Metric) Sample:
10 μL
15 μL
20 μL
25 μL
35 μL
45 μL
50 μL
65 μL
90 μL
Width (Metric) Teeth:
2.5 mm
4 mm
5.5 mm
9.25 mm
Unit Size:
Each
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Product Code. 11796185
missing translation for 'mfr': Fisherbrand™ TVGC1520
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
For use with TVG-SYS Vari-gel MIDI Horizontal Gel Electrophoresis Unit
  • Available in 1, 1.5 and 2mm thickness
Specifications
For Use With (Equipment) TVG-SYS Vari-Gel MIDI Horizontal Gel Electrophoresis Unit
Type Comb
No. of Wells 20
Thickness (Metric) 1.5 mm
Volume (Metric) Sample 15 μL
Width (Metric) Teeth 2.5 mm