Learn More
Fisherbrand™ Combs for TV100 Standard Twin-Plate Mini-Gel Vertical Electrophoresis Unit
Click to view available options
No. of Wells:
10
12
Thickness (Metric):
0.75 mm
2 mm
Width (Metric) Teeth:
3.75 mm
4 mm
Volume (Metric) Well:
42 μL
112 μL
120 μL
Unit Size:
Each
Description
- Available in 0.75, 1, 1.5 and 2mm thickness
Specifications
Specifications
| For Use With (Equipment) | TV100 Standard Twin-Plate Mini-Gel Vertical Electrophoresis Unit |
| Type | Comb |
| No. of Wells | 12 |
| Thickness (Metric) | 0.75 mm |
| Volume (Metric) Sample | 42 μL |
| Volume (Metric) Well | 42 μL |
| Width (Metric) Teeth | 3.75 mm |
| Height (Metric) | Teeth: 15 mm |