Learn More
Fisherbrand™ Combs for TV100 Standard Twin-Plate Mini-Gel Vertical Electrophoresis Unit
Product Code. 10121611
Click to view available options
No. of Wells:
10
12
Thickness (Metric):
0.75 mm
2 mm
Width (Metric) Teeth:
3.75 mm
4 mm
Volume (Metric) Well:
42 μL
112 μL
120 μL
Unit Size:
Each
Product Code. 10121611
Brand: Fisherbrand™ TV100C210
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with TV100 Standard Twin-Plate Mini-Gel Vertical Electrophoresis Unit
  • Available in 0.75, 1, 1.5 and 2mm thickness
Specifications
For Use With (Equipment) TV100 Standard Twin-Plate Mini-Gel Vertical Electrophoresis Unit
Type Comb
No. of Wells 10
Thickness (Metric) 2 mm
Volume (Metric) Well 120 μL
Width (Metric) Teeth 4 mm
Height (Metric) Teeth: 15 mm