Learn More
Fisherbrand™ Combs for HU13 Midi Horizontal Gel Electrophoresis Unit, 2mm thick
Product Code. 10641702
Click to view available options
No. of Wells:
10
12
16
24
28
Width (Metric) Teeth:
3 mm
5.5 mm
8 mm
9.5 mm
Volume (Metric) Sample:
25 μL
50 μL
70 μL
85 μL
Unit Size:
Each
Product Code. 10641702
Brand: Fisherbrand™ HU13C216
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with HU13 Midi Horizontal Gel Electrophoresis Unit
  • Color coded
  • Height adjustable
  • Offer complete control over loading volume
  • Well depth to maximum 112-sample throughput
Specifications
For Use With (Equipment) HU13 Midi Horizontal Gel Electrophoresis Unit
Type Comb
No. of Wells 16
Thickness (Metric) 2 mm
Volume (Metric) Sample 50 μL
Width (Metric) Teeth 5.5 mm