missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 1.5mm Thick

Product Code. 11592884 Shop All Fisherbrand Products
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
16
2 Prep, 2 Marker
20
25
4 Prep, 2 Marker
8
Volume (Metric) Sample:
15 μL
20 μL
24 μL
34 μL
41 μL
61 μL
115 μL
236 μL
540 μL
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Volume (Metric) Sample unitSize
11592884 10 41 μL Each
11813313 10 41 μL Each
11843313 1 Prep, 1 Marker 540 μL Each
11883313 12 34 μL Each
11823323 16 24 μL Each
11873323 20 20 μL Each
11803333 2 Prep, 2 Marker 236 μL Each
11843333 25 15 μL Each
11883333 4 Prep, 2 Marker 115 μL Each
11823343 8 61 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 11592884 Supplier Fisherbrand™ Supplier No. 731034
This item is not returnable. View return policy

For use in preparatory electrophoresis and high-throughput screening.

  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximise sample volume
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10-sample and 20-sample combs are Multichannel pipettor compatible

Specifications

Color Red
For Use With (Equipment) Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit
Type Comb
No. of Wells 10
Quantity 1 Each
Thickness (Metric) 1.5 mm
Volume (Metric) Sample 41 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.