Learn More
Fisherbrand™ Combs for CHU25 Maxi-Plus Cooled Horizontal Gel Electrophoresis Unit
Product Code. p-8013121
Click to view available options
No. of Wells:
26
52
Thickness (Metric):
1 mm
1.5 mm
Width (Metric) Teeth:
3 mm
7 mm
Volume (Metric) Sample:
20 μL
30 μL
45 μL
Unit Size:
Each
Product Code. 10080181
Brand: Fisherbrand™ HU25C1.526MC
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with CHU25 Maxi-Plus Cooled Horizontal Gel Electrophoresis Unit
  • Color coded
  • Height adjustable
  • Offer complete control over loading volume and well depth
Specifications
For Use With (Equipment) CHU25 Maxi-Plus Cooled Horizontal Gel Electrophoresis Unit
Type Comb
No. of Wells 26
Thickness (Metric) 1.5 mm
Volume (Metric) Sample 45 μL
Width (Metric) Teeth 7 mm