Learn More
Copan Italia™ Combined Loop and Handle
Click to view available options
Diameter (Metric) Inside:
2 mm
3 mm
4 mm
5 mm
Unit Size:
Pack of 5
Specifications
Specifications
| Diameter (Metric) Inside | 4 mm |
| Product Type | Combined Loop and Handle |
| Gauge | 26 G |
| Quantity | 5 Pk. |
| Calibrated | No |