Learn More
Lenz Laborglasintrumente Columns Acc. to Hempel
Product Code. 10129310
Click to view available options
Standard Taper Joint:
Cone and Socket NS 14/23
Cone and Socket NS 24/29
Cone and Socket NS 29/32
Length (Metric):
300 mm
500 mm
800 mm
Unit Size:
Each
Product Code. 10129310
Brand: Lenz Laborglasintrumente 05115301
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Columns acc. to Hempel
  • With removable glass jacket
  • For packings like Raschig rings or glass beads
  • With NS socket and NS cone
  • With removable glass jacket
  • Made of DURAN™ tubing
Specifications
Length (Metric) 300 mm
Standard Taper Joint Cone and Socket NS 14/23