Learn More
Cytiva Adapter for XK Columns
Tienda Cytiva Productos
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
XK Empty Column, 16 mm Diameter
XK Empty Column, 26 mm Diameter
XK Empty Column, 50 mm Diameter
missing translation for 'unitSize'
Each
Descripción
XK empty columns are designed for liquid chromatography at low to medium pressure. They are user friendly, robust, and give reproducible packing, and they feature fittings for direct connection to ÄKTA™ design instruments or other HPLC systems. XK columns are supplied with a movable adapter.
- Plunger design ensures a homogeneous buffer distribution and reproducible packing
- QuickLock mechanism of the adapter shaft facilitates easy column handling and cleaning
- Adapter top-end cap fits directly into the column end piece
- Plunger carries the net ring and support screen for the medium bed
- An adjusting knob seals the adapter to the column wall
Especificaciones
Especificaciones
| Connection Type | 1.58 mm |
| For Use With (Application) | Liquid Chromatography at Low to Medium Pressure |
| For Use With (Equipment) | XK Empty Column, 26 mm Diameter |
| Type | Adapter for XK 26 Columns |
| Includes | Plunger, Top-end cap, Adjusting knob |
| Quantity | 1 Pack |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido