Learn More
SCAT Collectors for HPLC
Product Code. p-7237251 Shop All SCAT Europe Products
Click to view available options
Product Type:
2-in-1 Connector
3-in-1 Connector
3-in-1 Sideways Connector
3-in-1 Vertical Connector
8-in-1 Connector
Diameter (Metric) Inner:
5.0 to 11.5 mm
Unit Size:
Each
Diameter (Metric) Outer:
3.2 mm
Product Code. 10588140
Brand: SCAT 160139
Only null left
Add to basket
Add to basket
Collect several tubes into a single connector - no problem with the right collector! The tube connector is the most flexible connector of the SCAT system.
For a reliable and clean connection of hoses and capillaries for collecting liquids. SCAT collectors are available in versions with up to 8 connection options.The connections are either on the side or vertically, so that the tubes can be connected easily. Tubes with different diameters from 2.3 to 11.5 mm can be connected. The connectors are made of chemical-resistant PTFE, PFA or PP material.
Order Info
dimForUseWith
Specifications
Autoclavable Autoclavable
Diameter (Metric) Inner 5.0 to 11.5 mm
Material PTFE
For Use With Tube Connector
Shape Sideways
Includes Fittings for 2 x 5.0 to 11.5 mm ID, 2-in-1 Connector
Color White
Product Type 2-in-1 Connector