Learn More
Colistin sodium methanesulfonate, Thermo Scientific™
Brand:  Thermo Scientific Alfa Aesar J66698.06
Quantity 5g
 View more versions of this product
Product Code. 15428159
  • 633.00€ / 5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Freely soluble in water
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Identifiers
8068-28-8
1749.811
IQWHCHZFYPIVRV-KLWRPAJRSA-I
131668505
CCC(C)CCCCC(=NC(CCNCS(=O)(=O)O)C(=O)NC(C(C)O)C(=O)NC(CCNCS(=O)(=O)O)C(=NC1CCNC(=O)C(NC(=O)C(N=C(C(NC(=O)C(NC(=O)C(N=C(C(N=C1[O-])CCNCS(=O)(=O)O)[O-])CC(C)C)CC(C)C)CCNCS(=O)(=O)O)[O-])CCNCS(=O)(=O)O)C(C)O)[O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+]
C58H105N16Na5O28S5
MFCD00130824
Colimycin sodium methanesulfonate; Polymyxin E sodium methanesulfonate
pentasodium;(6S)-N-[(2R)-1-[[(2R,3S)-3-hydroxy-1-[[(2S)-1-[[(3S,6S,9S,12S,15R,18S,21S)-3-[(1R)-1-hydroxyethyl]-12,15-bis(2-methylpropyl)-8,17,20-trioxido-2,5,11,14-tetraoxo-6,9,18-tris[2-(sulfomethylamino)ethyl]-1,4,7,10,13,16,19-heptazacyclotricosa-7,16,
Specifications
8068-28-8
5g
MFCD00130824
Colimycin sodium methanesulfonate; Polymyxin E sodium methanesulfonate
IQWHCHZFYPIVRV-KLWRPAJRSA-I
pentasodium;(6S)-N-[(2R)-1-[[(2R,3S)-3-hydroxy-1-[[(2S)-1-[[(3S,6S,9S,12S,15R,18S,21S)-3-[(1R)-1-hydroxyethyl]-12,15-bis(2-methylpropyl)-8,17,20-trioxido-2,5,11,14-tetraoxo-6,9,18-tris[2-(sulfomethylamino)ethyl]-1,4,7,10,13,16,19-heptazacyclotricosa-7,16,
131668505
Colistin sodium methanesulfonate
White to Yellow
C58H105N16Na5O28S5
14,2479
Freely soluble in water
CCC(C)CCCCC(=NC(CCNCS(=O)(=O)O)C(=O)NC(C(C)O)C(=O)NC(CCNCS(=O)(=O)O)C(=NC1CCNC(=O)C(NC(=O)C(N=C(C(NC(=O)C(NC(=O)C(N=C(C(N=C1[O-])CCNCS(=O)(=O)O)[O-])CC(C)C)CC(C)C)CCNCS(=O)(=O)O)[O-])CCNCS(=O)(=O)O)C(C)O)[O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+]
1749.811
1759.89
Product Suggestions
Product Suggestions
Videos
SDS Documents