missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ COL4A6 (Human) Recombinant Protein
Human COL4A6 full-length ORF ( AAH05305, 23 a.a. - 73 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001288-P02.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: GEKSYGKPCGGQDCSGSCQCFPEKGARHNLQLLNDMAGRLYHFSEVLPNLF
Specifications
AAH05305 | |
collagen, type IV, alpha 6 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COL4A6 | |
GST |
1288 | |
Wheat germ expression system | |
25 μg | |
MGC88184 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |