missing translation for 'onlineSavingsMsg'
Learn More

Coenzyme A Trilithium Salt Dihydrate ≥96%, White Crystalline Powder, MP Biomedicals™

Product Code. 11427550 Shop All MP Biomedical Products
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
250 mg
500 mg
Unit Size:
100mg
10mg
250mg
25mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 11427550

Brand: MP Biomedicals 0210049390

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 18439-24-2
Molecular Formula C21H33Li3N7O16P3S
Molecular Weight (g/mol) 785.329
MDL Number MFCD00075848
InChI Key QSCBPHBAFBVXRK-UHFFFAOYSA-K
Synonym coenzyme a, trilithium salt
PubChem CID 53442190
IUPAC Name trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate
SMILES [Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O

Specifications

Color White
Assay Percent Range ≥96%
Quantity 500 mg
Formula Weight 821.4
Physical Form Crystalline Powder
Chemical Name or Material Coenzyme A Trilithium Salt Dihydrate >96%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.