Learn More
behr Labor-Technik™ COD Digestor System
Product Code. 10074130
Change view
Click to view available options
No. of Places:
8
12
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Places unitSize
10074130 8 Each
10772001 12 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10074130 Supplier behr Labor-Technik™ Supplier No. B00218405
  • ONLINE EXCLUSIVE PRICE
    7523.25€
    5151.33€ / Pack of 1
    Save 2371.92€
    31% Off
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Capable of simultaneous EPA, ISO, or DIN method digestion of up to 8/12 COD samples. behr Labor-Technik™ COD Digestor System is a workstation comprising all necessary components for COD sample digestion.
  • Systems consist of the following components:
    • Microprocessor-controlled TRS 200 time/temperature regulator with COD program
    • Precision heating block CSB/E for RG2 vessels
    • Holding frame E/B for RG2 sample vessels
    • Cooling bath KW/N with mounting for E/B frame
    • Sample vessels RG 2
    • Air-cooled condensers LK 1
    • Condenser stand LS for LK condensers
Specifications
Product Type COD Digestor
Includes Time/temperature regulator, heating block, holding frame, cooling bath, sample vessels, air-cooled condensers, condenser stand
No. of Places 8