Learn More
KARTELL COBAS MIRA™ Multi-Curvette
Product Code. p-200004217
Click to view available options
Quantity:
1 Segment
15 Segments
Unit Size:
Pack of 450
Product Code. 10767524
Brand: KARTELL 0150100
The COBAS MIRA™ Multi-Curvette is a 12-position polymethyl methacrylate (PMMA) multiple curvette segement for the Cobas Mira™ analyzer.
  • 12-position polymethyl methacrylate (PMMA) multiple cuvette segments for the Cobas Mira™; analyzer.
  • A high-tech production process and stringent quality control guarantee high reproducibility.
  • Available in racks of 15 cuvette segments for devices with continuous throughput or in single segments.
Specifications
Type Multi-cuvette
Sample Type Serum, Plasma, Urine, Whole Blood
Quantity 1 Segment