missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ YNB, Powdered Media: Without Ammonium Sulfate/Folic Acid/Riboflavin
Shop All MP Biomedical Products
Click to view available options
:
Without ammonium sulfate/folic acid/riboflavin; 227g
Description
- Nitrogen-based yeast
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Specifications
| Certifications/Compliance | Molecular Biology Certified™ |
| Content And Storage | 15°C to 30°C |
| Form | Powder |
| Product Type | Microbiology Media |
| Formulation | Trace Elements: Boric Acid 0.5mg/L, Copper Sulfate 0.04mg/L, Potassium Iodide 0.1mg/L, Ferric Chloride 0.2mg/L, Manganese Sulfate 0.4mg/L, Sodium Molybdate 0.2mg/L, Zinc Sulfate 0.4mg/L, Salts: Magnesium Sulfate 500mg/L, Sodium Chloride 100mg/L, Potassium Phosphate monobasic 1000mg, Calcium Chloride 132mg; Vitamins: 4-Amino Benzoic Acid 0.2mg, Biotin 0.002mg, Inositol 2mg, Nicotinic Acid 0.4mg, Panthotenic Acid Hemi Calcium Salt 0.4mg, Pyridoxine Hydrochloride 0.4mg, Thiamine Hydrochloride 0.4mg |
| For Use With (Application) | For vigorous growth of Saccharomyces cerevisiae |
| Quantity | 227 g |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur