Learn More
Cobalt Carbonyl (Stabilized with 1-5% Hexane), TRC
Specifications
Specifications
| Chemical Name or Material | Cobalt Carbonyl |
| CAS | 10210-68-1 |
| Molecular Formula | C8Co2O8 |
| Quantity | 1 g |
| Synonym | Cobalt octacarbonyl (Co2(CO)8), Cobalt Tetracarbonyl Dimer, Di-μ-carbonylhexacarbonyldicobalt, Dicobalt Carbonyl, Dicobalt Octacarbonyl, Octacarbonyldicobalt |
| InChI Formula | InChI=1S/8CO.2Co/c8*1-2;;/q;;;;;;;;2*+2 |
| SMILES | [Co+2].[Co+2].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+] |
| IUPAC Name | carbon monoxide;cobalt(2+) |
| Molecular Weight (g/mol) | 341.95 |
| Formula Weight | 341.8257 |