Learn More
Buerkle™ Thread adapters LaboPlastBuerkle™ 3/4 in.
Product Code. p-8003060 Shop All Bürkle Products
Click to view available options
Thread Size:
3/4 in. Captive Cap
3/4 in. Inflowing, 2 in. Mauser Outflowing
3/4 in. Inflowing, 2 in. Tri-Sure Outflowing
3/4 in. Inflowing, Conical R 3/4 in. Outflowing
Cylindrical G 3/4 in. External, 3/4 in. Internal
Fine Thread 2 in. External, 3/4 in. Internal
Hunersdorff External, 3/4 in. Internal
Kautex External, 3/4 in. Internal
Color:
Black
Blue
Gray
Green
Green/White
Red
Unit Size:
Each
Product Code. 10030042
Brand: Buerkle™ 06000120
Only null left
Add to basket
Add to basket
StopCock spigots, ball valves and other accessories can be connected to most commercially available containers, with the LaboPlast™ thread adapters.
  • All adapters have a standard outer thread R 3/4 in.
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene, Polyethylene
Thread Size 3/4 in. Inflowing, 2 in. Mauser Outflowing
Color Green/White
Product Type Thread Adapter