Learn More
Buerkle™ Thread adapters LaboPlastBuerkle™ 3/4 in.
StopCock spigots, ball valves and other accessories can be connected to most commercially available containers, with the LaboPlast™ thread adapters.
Brand: Buerkle™ 0600-0190
17.36 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.03800kg
Description
- All adapters have a standard outer thread R 3/4 in.
Specifications
Thread Adapter | |
Polypropylene | |
Fine thread 2 in. external; 3/4 in. internal | |
Green/White |