Learn More
3M™ Coarse Anti-Slip Tape
Description
- Anti-slip tape for extreme and heavy traffic areas
- Extra-large abrasive particles resist clogging in muddy or oily conditions
- Can be painted without losing friction properties
- Suitable for use on agriculture and forestry equipment, road building, civil engineering, lorries, tractors, construction vehicles, cranes and building equipment
- High performance adhesive
Specifications
Specifications
| Length (Metric) | 20 m |
| Packaging Quantity | 1/Cs. |
| Width (Metric) | 50 mm |
| Color | Black |