missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor III/Tissue Factor Rat anti-Mouse, Clone: 355220, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3178-SP
This item is not returnable.
View return policy
Description
Coagulation Factor III/Tissue Factor Monoclonal specifically detects Coagulation Factor III/Tissue Factor in Mouse samples. It is validated for Western Blot.Specifications
| Coagulation Factor III/Tissue Factor | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P20352 | |
| F3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Coagulation Factor III Ala29-Glu251 Accession # P20352 | |
| 25 μg | |
| Primary | |
| Detects mouse Coagulation Factor III/Tissue Factor in direct ELISAs and Western blots. | |
| Mouse | |
| Purified |
| Western Blot | |
| 355220 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), F3, FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2152 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2c |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction