Learn More
Description
Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.
High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.
Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.
High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.
Features
- Convenient stand-alone enzymes and buffers for workflow optimization
- No animal-origin components in final product composition
- Reliable and reproducible results with rigorous quality testing
- Scalable and available as OEM products
Applications
- CRISPR/Cas-based genome editing
- Gene expression
- Generation of pluripotent cells
Specifications
Specifications
| Chemical Name or Material | Manganese Choride |
| Content And Storage | 100 mM MnCl2, 250 μL Store at -20°C. |
| Concentration | 100 mM |
| Format | Vial |
| No. of Reactions | 100 Reactions |
| Molecular Formula | MnCl2 |
| Product Type | Polymerase Co-factor |
| Quantity | 250 μL |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.