missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Clozapine N-oxide dihydrochloride
GREENER_CHOICE

Product Code. p-7116123
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18776336

Brand: Tocris Bioscience™ 6329/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Activator of muscarinic DREADDs; water soluble version of Clozapine N-oxide (Cat. No. 4936)

Clozapine N-oxide dihydrochloride is a water-soluble salt of Clozapine N-oxide (Cat. No.4936). Activator of muscarinic DREADDs. Clozapine N-oxide dihydrochloride exhibits improved bioavailability (6 - 7-fold higher plasma concentration) compared with CNO, with less conversion to clozapine in animal studies.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Chloro-11-(4-methyl-4-oxido-1-piperazinyl)-5 H-dibenzo[b,e][1,4 ]diazepine dihydrochloride
CAS 2250025-93-3
Assay Percent Range >99%
Quantity 10 mg
Target DREADD Activators
Molecular Formula C18H19ClN4O0.2 HCl
Formula Weight Observed MW: 415.74
Purity 99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.