missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cloxacillin sodium salt monohydrate

Product Code. 15565136 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15565136

Brand: Thermo Scientific Chemicals 455300050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Cloxacillin sodium salt monohydrate, CAS # 7081-44-9, is a compound that targets gram-positive bacteria and shows antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Cloxacillin sodium salt monohydrate is a chlorinated derivative of oxacillin that shows β-lactamase-resistant antibiotic activity

• This compound inhibits the last stage of bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs), which results in cell lysis, that is mediated by bacterial cell wall autolytic enzymes

Applications

• Cloxacillin sodium salt monohydrate targets gram-positive bacteria, and is effective against staphylococci that produce β-lactamase

• It acts in vitro against both penicillin gram-resistant and gram-sensitive staphylococci, certain streptococci, and pneumococci

• This compound is commonly used in vivo and in vitro studies as AmpC β-lactamase inhibitor in combination with β-lactam antibiotics

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7081-44-9
Molecular Formula C19H19ClN3NaO6S
Molecular Weight (g/mol) 475.876
InChI Key KCUWTKOTPIUBRI-VICXVTCVSA-M
PubChem CID 23675743
ChEBI CHEBI:34978
IUPAC Name sodium;(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrate
SMILES CC1=C(C(=NO1)C2=CC=CC=C2Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)[O-].O.[Na+]

Specifications

Type Cloxacillin Sodium Salt Monohydrate
Infrared Spectrum Authentic
Water 3.0 to 5.0%
Quantity 5 g
Formula Weight 475.88
Percent Purity ≥94% (HPLC)
Chemical Name or Material Cloxacillin sodium salt monohydrate
compliance-icons compliance-icons
Product Identifier
  • Cloxacillin sodium salt monohydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.