missing translation for 'onlineSavingsMsg'
Learn More

Clove Oil, Fisher Chemical

Product Code. 10459550 Shop All Fisher Chemical Products
Click to view available options
:
100mL, Amber glass bottle
This item is not returnable. View return policy

Product Code. 10459550

Brand: Fisher Chemical C/6280/05

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

100mL, Amber glass bottle

TRUSTED_SUSTAINABILITY

Specifications

CAS 8000-34-8
Color Yellow
Boiling Point 250°C
Packaging Amber glass bottle
Molecular Formula C7H12ClN3O2
MDL Number 130815
Quantity 100 mL
InChI Key VEEIFXWJNCAVEQ-UHFFFAOYNA-N
Molecular Weight (g/mol) 205.64
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.