Learn More
Clopidol, TRC
Product Code. 30138481
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138481 10 g 10g
17962235 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30138481 Supplier Toronto Research Chemicals Supplier No. TRCC58728510G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2971-90-6
Molecular Formula C7 H7 Cl2 N O
Molecular Weight (g/mol) 192.04
Synonym 3,5-Dichloro-2,6-dimethyl-4-pyridinol, 2,6-Dimethyl-3,5-dichloro-4-hydroxypyridine, 2,6-Dimethyl-3,5-dichloro-4-pyridinol, 3,5-Dichloro-2,6-dimethyl-4-hydroxypyridine, 3,5-Dichloro-2,6-dimethylpyridinol, 3,5-Dichloro-4-hydroxy-2,6-dimethylpyridine, Clopidol, Coccidiostat C, Coyden, Coyden 25, Farmcoccid, Lerbek, Methylchloropindol, Methylchlorpindol, Meticlorpindol, NSC 253479, Pharmcoccid, Pharmcoccid 10, WR 61112
IUPAC Name 3,5-dichloro-2,6-dimethylpyridin-4-ol
SMILES Cc1nc(C)c(Cl)c(O)c1Cl
Specifications
Quantity 10 g
InChI Formula InChI=1S/C7H7Cl2NO/c1-3-5(8)7(11)6(9)4(2)10-3/h1-2H3,(H,10,11)
Formula Weight 190.9905
Percent Purity >95%
Chemical Name or Material Clopidol
Grade HPLC