Learn More
STEMCELL Technologies ClonaCell™-HY PEG
Click to view available options
Quantity:
1.5 mL
Unit Size:
1.50mL
Description
- ClonaCell™-HY PEG is suitable for inducing cellular fusion of myeloma cells with lymphocytes to produce hybridomas
- This reagent has been verified for use during mouse and rat hybridoma development and reportedly is compatible for production, cloning, and expansion of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster
- ClonaCell™-HY PEG is supplied as a 50% (w/v) solution of PEG 3350 in Dulbecco's Modified Eagle's Medium (DMEM) with gentamicin and phenol red
- The solution has been pre-tested and qualified for cell fusion
Specifications
Specifications
| Cell Type | Hybridomas |
| For Use With (Application) | Cell Culture, Hybridoma Generation |
| Quantity | 1.5 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.