Learn More
STEMCELL Technologies ClonaCell™-CHO ACF Supplement
Product Code. 15217678
Encompass
Click to view available options
Quantity:
2.5 mL
Unit Size:
2.5mL
Product Code. 15217678
Brand: STEMCELL Technologies 03820
Animal component-free, serum-free, glutamine-free 40X supplement for cloning during CHO cell line development
  • ClonaCell™-CHO ACF Supplement is a serum-free and animal component-free supplement that is added to protein-free media in order to increase cloning efficiency and promote robust growth of CHO cells
  • The 40X supplement can be added to a variety of protein-free liquid or semi-solid cell culture media to increase the growth and single-cell cloning efficiency of suspension-adapted CHO cells
  • The supplement contains recombinant proteins and phenol red, and does not contain animal components, serum, hydrolysates, L-glutamine, or selection agents
Specifications
Cell Line CHO Cells
For Use With (Application) Cell Culture
Quantity 2.5 mL
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.