missing translation for 'onlineSavingsMsg'
Learn More

Clomiphene citrate, Thermo Scientific™

A selective estrogen receptor modulator

71.40€ - 155.00€

Chemical Identifiers

CAS 50-41-9
Molecular Formula C32H36ClNO8
Molecular Weight (g/mol) 598.089
MDL Number MFCD00058322
InChI Key PYTMYKVIJXPNBD-BTKVJIOYSA-N
Synonym clomifene citrate, clomiphene citrate, clomid, cis-clomiphene citrate, clomiphene a citrate, zuclomid, chloramiphene, milophene, serophene, z-clomiphene citrate
PubChem CID 6420009
IUPAC Name 2-[4-[(E)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CCN(CC)CCOC1=CC=C(C=C1)C(=C(C2=CC=CC=C2)Cl)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15455839
SDS View Documents
Thermo Scientific Alfa Aesar
J63663.04
2g
71.40€
2g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15465839
SDS View Documents
Thermo Scientific Alfa Aesar
J63663.09
10g
155.00€
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Selective estrogen receptor modulator, which acts by inhibiting the action of estrogen on the pituitary. Clomiphene Citrate is an activator of PKC and Gonad-stimulating principle.

Solubility
Slightly soluble in water, methanol, chloroform (slightly), and DMSO (10 mM). Insoluble in ether.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

50-41-9
598.089
PYTMYKVIJXPNBD-BTKVJIOYSA-N
6420009
CCN(CC)CCOC1=CC=C(C=C1)C(=C(C2=CC=CC=C2)Cl)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O
C32H36ClNO8
MFCD00058322
clomifene citrate, clomiphene citrate, clomid, cis-clomiphene citrate, clomiphene a citrate, zuclomid, chloramiphene, milophene, serophene, z-clomiphene citrate
2-[4-[(E)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid

Specifications

50-41-9
MFCD00058322
clomifene citrate, clomiphene citrate, clomid, cis-clomiphene citrate, clomiphene a citrate, zuclomid, chloramiphene, milophene, serophene, z-clomiphene citrate
PYTMYKVIJXPNBD-BTKVJIOYSA-N
2-[4-[(E)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
6420009
Clomiphene citrate
C32H36ClNO8
14,2386
Slightly soluble in water,methanol,chloroform (slightly),and DMSO (10 mM). Insoluble in ether.
CCN(CC)CCOC1=CC=C(C=C1)C(=C(C2=CC=CC=C2)Cl)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O
598.089
598.08
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H360

missing translation for 'einecsNumber' : 200-035-3

missing translation for 'rtecsNumber' : YE0875000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.