missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Clobetasol propionate
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Quantity | 1 g |
| Solubility | Solubility aq. soln.: insoluble. |
| Melting Point | 191.0°C to 195.0°C |
| Formula Weight | 466.97 |
| InChI Key | CBGUOGMQLZIXBE-XGQKBEPLSA-N |
| SMILES | CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CCl |
| IUPAC Name | (1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-1-(2-chloroacetyl)-9b-fluoro-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl propanoate |
| Molecular Weight (g/mol) | 466.97 |
| Chemical Name or Material | Cloβsol propionate |
Sicurezza e movimentazione
Identificativo prodotto
- Clobetasol propionate
- Danger
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H360-May damage fertility or the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 21-Chloro-9-fluoro-11.beta.,17-dihydroxy-16.beta.-methylpregna-1,4-diene-3,20-dione 17-propionate
RUO – Research Use Only
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto