Learn More
Clinofibrate, TRC
Specifications
Specifications
| CAS | 30299-08-2 |
| Molecular Formula | C28H36O6 |
| Quantity | 10 mg |
| Synonym | 1,1-Bis[4'-(1''-carboxy-1''-methylpropoxy)phenyl]cyclohexane, 2,2'-[Cyclohexylidenebis(4,1-phenyleneoxy)]bis[2-methyl-butanoic Acid, Lipoclin, S 8527 |
| InChI Formula | InChI=1S/C28H36O6/c1-5-26(3,24(29)30)33-22-14-10-20(11-15-22)28(18-8-7-9-19-28)21-12-16-23(17-13-21)34-27(4,6-2)25(31)32/h10-17H,5-9,18-19H2,1-4H3,(H,29,30)(H,31,32) |
| SMILES | CCC(C)(Oc1ccc(cc1)C2(CCCCC2)c3ccc(OC(C)(CC)C(=O)O)cc3)C(=O)O |
| IUPAC Name | 2-[4-[1-[4-(2-carboxybutan-2-yloxy)phenyl]cyclohexyl]phenoxy]-2-methylbutanoic acid |
| Molecular Weight (g/mol) | 468.58 |
| Formula Weight | 468.2512 |
| Chemical Name or Material | Clinofibrate |