Learn More
Thermo Scientific™ CLINIConic™ Rotor Adapters
Product Code. 10461185 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
10 mL Round-bottom Tube
5 mL Round-bottom or Blood Collection Tube
7 mL Round-bottom or Blood Collection Tube
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
10461185 5 mL Round-bottom or Blood Collection Tube Each
10581845 7 mL Round-bottom or Blood Collection Tube Each
10073054 10 mL Round-bottom Tube Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10461185 Supplier Thermo Scientific™ Supplier No. 11172595
Only null left
Add to basket
Add to basket
Incorporate these rotor adapters with the CLINIConic fixed-angle rotor.
Incorporate Thermo Scientific™ CLINIConic™ Rotor Adapters with the CLINIConic fixed-angle rotor.
Ordering Information:
CLINIConic fixed-angle rotor body (75-003-623) is available separately. Contact your sales representative for more information.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Tube Adapter
Tube Type 5 mL Round Bottom, Blood Collection Tube
Capacity 5 mL Round-bottom or Blood Collection Tube