Learn More
Nickel Electro™ Clifton™ Stainless Steel Basket
Product Code. p-8012482
Change view
Click to view available options
Capacity (Metric):
1 L
3 L
4 L
6 L
8 L
12 L
14 L
22 L
Dimensions (L x W x H):
177 x 73 x 30 mm
198 x 106 x 50 mm
241 x 260 x 95 mm
250 x 115 x 75 mm
212 x 185 x 110 mm
228 x 107 x 110 mm
250 x 190 x 115 mm
410 x 246 x 110 mm
455 x 250 x 115 mm
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Dimensions (L x W x H) unitSize
13101303 14 L 241 x 260 x 95 mm Each
13111303 22 L 410 x 246 x 110 mm Each
13191293 8 L 212 x 185 x 110 mm Each
13191313 1 L 177 x 73 x 30 mm Each
13101323 3 L 198 x 106 x 50 mm Each
13131323 12 L 455 x 250 x 115 mm Each
13111323 6 L 250 x 115 x 75 mm Each
13121323 12 L 250 x 190 x 115 mm Each
13181293 4 L 228 x 107 x 110 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 13101303 Supplier Nickel Electro™ Supplier No. BSK14
Only null left
Add to basket
Add to basket
Complete an Ultrasonic Bath with a stainless steel basket. Nickel Electro™ Clifton™ Stainless Steel Basket is durable and made of high quality stainless steel.
  • High quality
  • Durable
  • Stainless steel
  • For use with Ultrasonic Bath
  • Includes side lifting handles
Specifications
Type Basket
Capacity (Metric) 14 L
Depth (Metric) 260 mm
For Use With (Application) For Ultrasonic Bath
Height (Metric) 95 mm
Material Stainless Steel
Weight (Metric) 1.25 kg
Width (Metric) 241 mm
Dimensions (L x W x H) 241 x 260 x 95 mm