Learn More
Nickel Electro™ Clifton™ Bath Lab Armor™
Product Code. 12789008
Click to view available options
Capacity (Metric):
0.5 L
4 L
Unit Size:
Each
Product Code. 12789008
Brand: Nickel Electro™ BA0.5
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Obtain superior water bath cleaning results when you replace water in your water bath with these thermally efficient pellets that are easy to keep clean. This Clifton™ Bath Armor™ tackles water contamination and protects your water bath from drying out.
  • Tackles water contamination by utilizing a dry, metallic thermal media comprised of small beads instead of water
  • Thermally efficient pellets that are easy to keep clean
  • Moisture and gas impermeable
  • High thermal conductivity
  • Smooth and rounded almost fluid like
  • Reduces water consumption
  • Saves the need for racks, floaties and bottle weights
  • Do not autoclave
Specifications
Capacity (Metric) 0.5 L
Type Bath Lab Armor
Quantity 0.5 L