Learn More
Clidinium Bromide, TRC
Product Code. 30135917
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135917 1 g 1g
17914115 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30135917 Supplier Toronto Research Chemicals Supplier No. TRCC5762501G
 Restricted Item
Estimated Shipment: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3485-62-9
Molecular Formula C22 H26 N O3 . Br
Molecular Weight (g/mol) 432.35
Synonym Clidinium Bromide, 1-Azoniabicyclo[2.2.2]octane, 3-[(2-hydroxy-2,2-diphenylacetyl)oxy]-1-methyl-, bromide (1:1), 1-Azoniabicyclo[2.2.2]octane, 3-[(hydroxydiphenylacetyl)oxy]-1-methyl-, bromide (9CI), 3-Hydroxy-1-methylquinuclidinium bromide, benzilate (6CI,7CI), Quinuclidinium, 3-hydroxy-1-methyl-, bromide, benzilate (8CI), 1-Methyl-3-(benziloyloxy)quinuclidinium bromide, 3-(Benziloyloxy)-1-methylquinuclidinium bromide, Clidinium bromide, Quarzan, Quarzan bromide, Ro 2-3773
IUPAC Name (1-methyl-1-azoniabicyclo[2.2.2]octan-3-yl) 2-hydroxy-2,2-diphenylacetate;bromide
SMILES [Br-].C[N+]12CCC(CC1)C(C2)OC(=O)C(O)(c3ccccc3)c4ccccc4
Specifications
Quantity 1 g
InChI Formula InChI=1S/C22H26NO3.BrH/c1-23-14-12-17(13-15-23)20(16-23)26-21(24)22(25,18-8-4-2-5-9-18)19-10-6-3-7-11-19;/h2-11,17,20,25H,12-16H2,1H3;1H/q+1;/p-1
Formula Weight 431.1096
Chemical Name or Material Clidinium Bromide