missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Click N-Acetylmuramic acid - azide

Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
Click N-Acetylmuramic acid-azide is a derivative of the N-acetylmuramic acid (NAM) component of bacterial peptidoglycans. Incorporated into bacterial peptidoglycans during biosynthesis. Suitable for fluorescent labeling of peptidoglycans when ''click'-conjugated to a fluorescent dye.
Specifications
Specifications
| Chemical Name or Material | (2 R)-2-(((3 R,4 R,5 S,6 R)-3-(2-Azidoacetamido)-2,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2 H-pyran-4-yl)oxy)propanoic acid |
| CAS | 2245794-64-1 |
| Quantity | 2 mg |
| Target | Glycobiology Probes |
| Molecular Formula | C11H18N4O8 |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction