Learn More
Cletoquine-d4 Oxalate, TRC
Product Code. 30356145
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30356145 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30356145 Supplier Toronto Research Chemicals Supplier No. TRCC57350210MG
 Restricted Item
Estimated Shipment: 10-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Cletoquine-d4 Oxalate
CAS 1216461-57-2
Molecular Formula C18H20D4ClN3O5
Quantity 10 mg
Synonym 2-[[4-[(7-Chloroquinolin-4-yl)amino]-1,1,2,2-tetradeuteriopentyl]amino]ethanol oxalate, 2-[[4-[(7-Chloro-4-quinolinyl)amino]pentyl-1,1,2,2-d4]amino]ethanol ethanedioate (1:1), Cletoquine-d4 oxalate, Desethylhydroxychloroquine-d4 oxalate, Desethylhydroxychloroquine-D4 (1,4-diaminopentyl-1,1,2,2-D4) oxalate
InChI Formula InChI=1S/C16H22ClN3O.C2H2O4/c1-12(3-2-7-18-9-10-21)20-15-6-8-19-16-11-13(17)4-5-14(15)16;3-1(4)2(5)6/h4-6,8,11-12,18,21H,2-3,7,9-10H2,1H3,(H,19,20);(H,3,4)(H,5,6)/i2D2,7D2;
SMILES [2H]C([2H])(CC(C)Nc1ccnc2cc(Cl)ccc12)C([2H])([2H])NCCO.OC(=O)C(=O)O
IUPAC Name 2-[[4-[(7-chloroquinolin-4-yl)amino]-1,1,2,2-tetradeuteriopentyl]amino]ethanol;oxalic acid
Molecular Weight (g/mol) 401.88