Learn More
Epredia™ ClearVue™ Mountants
Product Code. 11376790 Shop All Epredia Products
Change view
Click to view available options
Solution Type:
Mountant, Toluene-based
Mountant, Xylene-based
Unit Size:
0.95 L
Each
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Solution Type Unit Size
11376790 Mountant, Toluene-based Each
11396790 Mountant, Xylene-based 0.95 L
12766788 Mountant, Xylene-based Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11376790 Supplier Epredia™ Supplier No. 4211
  • ONLINE EXCLUSIVE PRICE
    66.00€
    51.49€ / Pack of 1
    Save 14.51€
    21% Off
 Restricted Item
In Stock
Enjoy compatibility with a variety of automated coverslippers to accommodate any lab setting with Epredia™ ClearVue™ Mountants.
TRUSTED_SUSTAINABILITY
Specifications
Color Light yellow
Product Type Mountant
Quantity 1 pt.
Solution Type Mountant, Toluene-based
Formulation Toluene, Acrylic Resin
Form Liquid