Learn More
Diversey™ ClearKlens IPA VH1 Disinfectant
BIOPHARMA
Product Code. 17603245
Change view
Click to view available options
Volume (Metric):
500 mL
1 L
5 L
Description:
For AT, BE, CH, DE, FR, GB, IE, IT, NL only
For ES, PL, PT, RU only
For FI, NO, PL, PT, RU, SE only
Unit Size:
10 L
12 L
6 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Description unitSize
17603245 500 mL For ES, PL, PT, RU only 6 L
17613245 1 L For FI, NO, PL, PT, RU, SE only 12 L
17683095 5 L For AT, BE, CH, DE, FR, GB, IE, IT, NL only 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17603245 Supplier Diversey™ Supplier No. 101105066
Stock Up Your Lab
Use promo code 28007
View all promotions
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket
Ready to use disinfectant for non-porous hard surfaces and gloves
ClearKlens IPA is a non-sterile, ready to use disinfectant for the Life Sciences industry. It is used as general purpose mid-shift and terminal disinfectant. ClearKlens IPA is based on 70% isopropyl alcohol. ClearKlens IPA is available as 500 mL trigger spray, and as 1 L bottle or 5 L canister.
Key properties
  • ClearKlens IPA is a fast acting disinfectant spray for use in mid-shift and terminal disinfection applications.
  • It has a broad spectrum of efficacy against most vegetative forms of micro- organisms.
  • ClearKlens IPA is safe for use on food contact surfaces and evaporates rapidly, leaving no residue. It is suitable for the disinfection of alcohol-safe surfaces, machines and components (conveyors, fillers, bottle inspectors, water-sensitive components, etc.)
  • BPR approved pack and formulation
Benefits
  • Filtered at 0.2 μm
  • Ready to use
  • Effective broad-spectrum activity
  • Non foaming
  • Suitable for use in moisture sensitive areas
  • Readily evaporates leaving no residues
Specifications
Product Type Surface Disinfectant
pH 6 to 9
Sterility Non-sterile
Composition 70% IPA/30% deionized water
Container Type Spray Bottle
Description For ES, PL, PT, RU only
Quantity 12/Cs.
Volume (Metric) 500 mL
compliance-icons compliance-icons
Product Identifier
  • ClearKlens IPA VH1
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: propan-2-ol