Learn More
Buerkle™ Clear PVC Pressure Tubing
Product Code. 10048512 Shop All Bürkle Products
Change view
Click to view available options
Diameter (Metric) Inner:
8 mm
9 mm
10 mm
13.2 mm
Unit Size:
50m
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
10048512 9 mm 50m
10729694 10 mm 50m
10353273 13.2 mm 50m
10562954 8 mm 50m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
The pressure figures are maximum values (bursting pressure) under optimum ambient conditions. The user is responsible for determining any suitability!
Product Code. 10048512 Supplier Buerkle™ Supplier No. 88760915
Only null left
Add to basket
Add to basket
The pressure figures are maximum values (bursting pressure) under optimum ambient conditions. The user is responsible for determining any suitability!
Fabric reinforced, permanently transparent tubing for use with foodstuffs applications.
  • Increased flexibility
  • High chemical, ageing, and abrasion resistance
  • Environmentally friendly, cadmium-free
  • Foodstuffs quality acc. to the EC Guideline 1935/2004
  • Hardness Shore A 70 ± 3
  • Operating temperature of −20° to +60°C
  • Pressure figures are maximum values (bursting pressure) under optimum ambient conditions
  • TÜV-approved (German industry standards)
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EU Foodstuffs, TÜV-Approved
Diameter (Metric) Inner 9 mm
Material PVC
Diameter (Metric) Outer 15 mm
Overall Length (Metric) 50 m
Thickness (Metric) 3 mm
For Use With Foodstuff
For Use With (Application) Suitable for Use with Foodstuffs
Min. Operating Temperature (Metric) -20°C
Resistance Properties Abrasion-Resistant, Chemical-Resistant
Color Clear
Hardness 70 (Shore A, ± 3)
Maximum Pressure 16 bar
Max. Operating Temperature (Metric) 60°C
Product Type Pressure Tubing