Learn More
Berlin Packaging™ Clear Laboratory Glassware Pot
Product Code. 10195884
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Quantity:
18 Set
20 Set
48 Set
63 Set
Unit Size:
Pack of 2688
Pack of 4158
Pack of 48
Pack of 720
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Quantity unitSize
10195884 250 mL 48 Set Pack of 48
30408780 1000 mL 18 Set Pack of 720
30408786 100 mL 63 Set Pack of 4158
30408783 250 mL 48 Set Pack of 2688
30408784 500 mL 20 Set Pack of 720
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Product Code. 10195884 Supplier Berlin Packaging™ Supplier No. 1000721
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Use the Berlin Packaging™ Clear Laboratory Glassware Pot as a fundamental piece of laboratory glassware. The autoclavable clear glass is breakproof and chemically and thermally shockproof.
  • Clear borosilicate glass
  • Break-resistant
  • Chemically and thermally shockproof;
  • In compliance with EP, USP and JP guidelines
  • Autoclavable
Specifications
Certifications/Compliance DIN 55
Quantity 48 Set
Capacity (Metric) 250 mL
Material Glass
Product Type Pot