Learn More
Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
BIOPHARMA
Product Code. 10547915
Change view
Click to view available options
Surface Treatment:
Enhanced Binding
Immulon 1 B
Immulon 2 HB
Immulon 4 HBX
MaxiSorp
MediSorp
MultiSorp
PolySorp
Universal Binding
Certified:
No
Yes
Volume (Metric) Working:
230 μL
280 μL
330 μL
350 μL
Barcode:
Includes Barcode
No Barcode
Unit Size:
Pack of 100
Pack of 180
Pack of 50
Pack of 60
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Surface Treatment Certified Volume (Metric) Working Barcode Unit Size
10547915 Immulon 1 B No 230 μL No Barcode Pack of 50
10400482 Immulon 1 B No 230 μL No Barcode Pack of 100
10795026 Immulon 2 HB No 280 μL No Barcode Pack of 50
10468075 Immulon 4 HBX No 280 μL No Barcode Pack of 50
10394751 MaxiSorp Yes 350 μL No Barcode Pack of 60
10547781 MaxiSorp No 350 μL No Barcode Pack of 60
10498802 PolySorp No 350 μL No Barcode Pack of 180
10294401 MaxiSorp Yes 350 μL No Barcode Pack of 180
10264451 MaxiSorp Yes 350 μL Includes Barcode Pack of 180
10275411 MediSorp Yes 350 μL No Barcode Pack of 60
10565131 MultiSorp No 350 μL No Barcode Pack of 60
10449672 PolySorp No 350 μL No Barcode Pack of 60
10190825 Universal Binding No 330 μL No Barcode Pack of 50
10130665 Enhanced Binding Yes 330 μL No Barcode Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Each lot is functionally tested for IgG binding.
Product Code. 10547915 Supplier Thermo Scientific™ Supplier No. 3355
  • ONLINE EXCLUSIVE PRICE
    163.89€
    150.01€ / Pack of 50
    Save 13.88€
    8% Off
In Stock
Add to basket
Add to basket
Bulk Quote Request
Each lot is functionally tested for IgG binding.
Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.
Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
96-Well Solid Plates:
  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns
Surfaces:
  • Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.
Ordering Information:
Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Certified No
Format Standard
Detection Method Colorimetric
Array 8 x 12
Packaging 50 Cases
Binding Type Hydrophobic
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Volume (Metric) Working 230 μL
Sterility Non-sterile
Surface Treatment Immulon 1 B
Well Shape Round Bottom (U-well)
No. per Case 50
No. per Pack 10
Plate Blocking None
Target Molecule Lipids
Show More Show Less
For Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, is the surface treatment solely on the bottom of the well, or is it also applied to the sides?
For energy treated plates, the entire plate surface receives the energy treatment. The MaxiSorp, PolySorp, MediSorp and MultiSorp plates are energy treated plates. For traditionally treated plates, the well surface is treated up to the listed working volume.
Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?
Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.
For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?
The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.
Which Nunc immunodiagnostic passive surface should I choose: PolySorp, MediSorp, MaxiSorp, or MultiSorp?
Our passive immunodiagnostic surfaces are based on a series of treatments that modify the polystyrene surface to different final levels of hydrophilicity. Selection of one of these surfaces for ELISA development should be based on the characteristics of the biomolecule you seek to immobilize. PolySorp is fairly hydrophobic and should be used for biomolecules that have hydrophobic domains, e.g., lipids, lipoproteins, and large proteins. MediSorp is slightly hydrophilic and can be used for biomolecules with hydrophilic and hydrophobic properties, e.g., medium-to-large proteins such as albumin, or amphiphilic biomolecules such as LPS. MaxiSorp is quite hydrophilic, optimized for binding of IgG (and any other molecule with similar characteristics) and thus is a popular choice for sandwich ELISA and similar protocols. MultiSorp is very hydrophilic, and in turn will immobilize very hydrophilic biomolecules such as glycoproteins. Some optimization will always be necessary, and in many cases screening for binding optimization may involve a few surface types.
What is the difference between a certified and an uncertified immunodiagnostic plate, e.g., Cat. No. 439454 vs. 442404?
There is no difference in the manufacturing process. Certified plates are performance tested (using an antibody binding protocol) during quality control, whereas uncertified plates are not. This test is expensive for us to perform, so we offer the uncertified version at cost savings customers for whom antibody binding testing results are not critical.