Learn More
Greiner Bio-One μClear™ Bottom 96-well Polystyrene Microplates
Product Code. 10030262
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10030262 Black Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10030262 Supplier Greiner Bio-One Supplier No. 655096
  • ONLINE EXCLUSIVE PRICE
    454.96€
    271.55€ / Pack of 40
    Save 183.41€
    40% Off
Estimated Shipment: 14-12-2026
Minimises the risk of sample carryover and cross-contamination. Greiner Bio-One μClear™ Bottom 96-Well Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
  • Non-Pyrogenic
  • Material: Polystyrene
  • Free of detectable DNase, RNase, human DNA
  • Traced way back to production through defined LOT number system
  • More suitable for non polar proteins and peptides
  • Medium binding
Certifications
DIN ISO 9001
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type Medium Binding
Color Black
Product Type Assay Microplate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Medium Binding
Well Shape Flat Bottom (F-well)
Quantity 40/Cs.
Well Design Chimney