Learn More
CleanSpace Technology™ CleanSpace3™ Cleaning and Storage Plug Set
Product Code. 15970042 Shop All CleanSpace Technology Products
Change view
Click to view available options
For Use With (Equipment):
CleanSpace™ HALO Respirator unit (CS3000)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15970042 CleanSpace™ HALO Respirator unit (CS3000) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Do not wear the respirator when the plug set is in place.
Product Code. 15970042 Supplier CleanSpace Technology™ Supplier No. CS3011
  • ONLINE EXCLUSIVE PRICE
    66.00€
    54.50€ / Pack of 1
    Save 11.50€
    17% Off
Estimated Shipment: 12-10-2026
Do not wear the respirator when the plug set is in place.
Tool for use with the CleanSpace™ HALO Respirator unit (CS3000). Seals all the openings on the CleanSpace™ HALO PAPR unit for storage. Made of polycarbonate and silicone.
  • Used with the revolutionary CleanSpace HALO PAPR: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement
Specifications
Model CS3011
For Use With (Equipment) CleanSpace™ HALO Respirator unit (CS3000)